Merck
  • Persistent Interneuronopathy in the Prefrontal Cortex of Young Adult Offspring Exposed to Ethanol In Utero.
Persistent Interneuronopathy in the Prefrontal Cortex of Young Adult Offspring Exposed to Ethanol In Utero.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2015-08-08)
Alexander G J Skorput, Vivek P Gupta, Pamela W L Yeh, Hermes H Yeh
ABSTRACT
Gestational exposure to ethanol has been reported to alter the disposition of tangentially migrating GABAergic cortical interneurons, but much remains to be elucidated. Here we first established the migration of interneurons as a proximal target of ethanol by limiting ethanol exposure in utero to the gestational window when tangential migration is at its height. We then asked whether the aberrant tangential migration of GABAergic interneurons persisted as an enduring interneuronopathy in the medial prefrontal cortex (mPFC) later in the life of offspring prenatally exposed to ethanol. Time pregnant mice with Nkx2.1Cre/Ai14 embryos harboring tdTomato-fluorescent medial ganglionic eminence (MGE)-derived cortical GABAergic interneurons were subjected to a 3 day binge-type 5% w/w ethanol consumption regimen from embryonic day (E) 13.5-16.5, spanning the peak of corticopetal interneuron migration in the fetal brain. Our binge-type regimen increased the density of MGE-derived interneurons in the E16.5 mPFC. In young adult offspring exposed to ethanol in utero, this effect persisted as an increase in the number of mPFC layer V parvalbumin-immunopositive interneurons. Commensurately, patch-clamp recording in mPFC layer V pyramidal neurons uncovered enhanced GABA-mediated spontaneous and evoked synaptic transmission, shifting the inhibitory/excitatory balance toward favoring inhibition. Furthermore, young adult offspring exposed to the 3 day binge-type ethanol regimen exhibited impaired reversal learning in a modified Barnes maze, indicative of decreased PFC-dependent behavioral flexibility, and heightened locomotor activity in an open field arena. Our findings underscore that aberrant neuronal migration, inhibitory/excitatory imbalance, and thus interneuronopathy contribute to indelible abnormal cortical circuit form and function in fetal alcohol spectrum disorders. The significance of this study is twofold. First, we demonstrate that a time-delimited binge-type ethanol exposure in utero during early gestation alters corticopetal tangential migration of GABAergic interneurons in the fetal brain. Second, our study is the first to integrate neuroanatomical, electrophysiological, and behavioral evidence that this "interneuronopathy" persists in the young adult offspring and contributes to enduring changes in (1) the distribution of parvalbumin-expressing GABAergic cortical interneurons in the medial prefrontal cortex, (2) GABA-mediated synaptic transmission that resulted in an inhibitory/excitatory synaptic imbalance, and (3) behavioral flexibility. These findings alert women of child-bearing age that fetal alcohol spectrum disorders can be rooted very early in fetal brain development, and reinforce evidence-based counseling against binge drinking even at the earliest stages of pregnancy.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O