Merck
  • N-Acetylneuraminic Acid Supplementation Prevents High Fat Diet-Induced Insulin Resistance in Rats through Transcriptional and Nontranscriptional Mechanisms.
N-Acetylneuraminic Acid Supplementation Prevents High Fat Diet-Induced Insulin Resistance in Rats through Transcriptional and Nontranscriptional Mechanisms.
BioMed research international (2015-12-22)
Zhang Yida, Mustapha Umar Imam, Maznah Ismail, Norsharina Ismail, Nur Hanisah Azmi, Waiteng Wong, Hadiza Altine Adamu, Nur Diyana Md Zamri, Aini Ideris, Maizaton Atmadini Abdullah
ABSTRACT
N-Acetylneuraminic acid (Neu5Ac) is a biomarker of cardiometabolic diseases. In the present study, we tested the hypothesis that dietary Neu5Ac may improve cardiometabolic indices. A high fat diet (HFD) + Neu5Ac (50 or 400 mg/kg BW/day) was fed to rats and compared with HFD + simvastatin (10 mg/kg BW/day) or HFD alone for 12 weeks. Weights and serum biochemicals (lipid profile, oral glucose tolerance test, leptin, adiponectin, and insulin) were measured, and mRNA levels of insulin signaling genes were determined. The results indicated that low and high doses of sialic acid (SA) improved metabolic indices, although only the oral glucose tolerance test, serum triglycerides, leptin, and adiponectin were significantly better than those in the HFD and HFD + simvastatin groups (P < 0.05). Furthermore, the results showed that only high-dose SA significantly affected the transcription of hepatic and adipose tissue insulin signaling genes. The data suggested that SA prevented HFD-induced insulin resistance in rats after 12 weeks of administration through nontranscriptionally mediated biochemical changes that may have differentially sialylated glycoprotein structures at a low dose. At higher doses, SA induced transcriptional regulation of insulin signaling genes. These effects suggest that low and high doses of SA may produce similar metabolic outcomes in relation to insulin sensitivity through multiple mechanisms. These findings are worth studying further.
Product Description
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Pricing and availability is not currently available.
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Pricing and availability is not currently available.
Supelco
D-(+)-Glucose, analytical standard
Pricing and availability is not currently available.
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Pricing and availability is not currently available.
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Pricing and availability is not currently available.
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Pricing and availability is not currently available.