Merck
  • Mammary Branching Morphogenesis Requires Reciprocal Signaling by Heparanase and MMP-14.
Mammary Branching Morphogenesis Requires Reciprocal Signaling by Heparanase and MMP-14.
Journal of cellular biochemistry (2015-03-05)
Angélica Maciel Gomes, Ramray Bhat, Ana Luísa Correia, Joni D Mott, Neta Ilan, Israel Vlodavsky, Mauro S G Pavão, Mina Bissell
ABSTRACT
The development of the mammary gland involves formation of a branched arboreal structure resulting from the penetration and proliferation of epithelial cells into the fat pad. The mammary cells invade by remodeling their surrounding extracellular matrix (ECM), which are rich in proteins, and glycans such as heparan sulfate proteoglycans (HSPGs). There is increasing literature on how the interaction between signaling by ECM and matrix metalloproteinases (MMPs) is relevant to morphogenetic and physiological contexts. Here we sought to understand how heparanase, the sole mammalian heparan sulfate-degrading endoglycosidase may regulate mammary gland development. We found a robust localization of heparanase within growing end buds during branching in vivo. Using three-dimensional (3D) organotypic cultures, we showed that heparanase expression and activity are required for mammary epithelial invasion/branching within dense collagen I gels. Morphometric analysis of glands from both heparanase-overexpressing and knockout mice showed a direct correlation between degree of branching and the heparanase levels, confirming our 3D organotypic culture observations. Finally, we uncovered a reciprocal association between levels of heparanase and MMP14, a membrane-bound MMP, shedding further light on how branching occurs within developing mammary glands.
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
SAFC
Sodium chloride solution, 5 M
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis