Merck
Photoinduced spectral changes of photoluminescent gold nanoclusters.
Journal of biomedical optics (2014-12-18)
Marija Matulionytė, Raminta Marcinonytė, Ričardas Rotomskis
ABSTRACT
Ultrasmall photoluminescent gold nanoclusters (Au NCs), composed of several atoms with sizes up to a few nanometers, have recently stimulated extensive interest. Unique molecule-like behaviors, low toxicity, and facile synthesis make photoluminescent Au NCs a very promising alternative to organic fluorophores and semiconductor quantum dots (QDs) in broad ranges of biomedical applications. However, using gold nanoparticles (Au NPs) for bioimaging might cause their degradation under continuous excitation with UV light, which might result in toxicity. We report spectral changes of photoluminescent 2-(N-morpholino) ethanesulfonic acid (MES)-coated (Au-MES) NCs under irradiation with UV/blue light. Photoluminescent water soluble Au- MES NCs with a photoluminescence (PL) band maximum at 476 nm (λex = 420 nm) were synthesized. Under irradiation with 402 nm wavelength light the size of photoluminescent Au-MES NCs decreased (λem = 430 nm). Irradiating the sample solution with 330 nm wavelength light, nonluminescent Au NPs were disrupted, and photoluminescent Au NCs (λem = 476 nm) were formed. Irradiation with 330 nm wavelength light did not directly affect photoluminescent Au-MES NCs, however, increase in PL intensity indicated the formation of photoluminescent Au NCs from the disrupted nonluminescent Au NPs. This study gives a good insight into the photostability of MES-coated Au NPs under continuous excitation with UV/blue light.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Molecular Biology, sterile filtered