Merck
  • Cyclin Y phosphorylation- and 14-3-3-binding-dependent activation of PCTAIRE-1/CDK16.
Cyclin Y phosphorylation- and 14-3-3-binding-dependent activation of PCTAIRE-1/CDK16.
The Biochemical journal (2015-07-25)
Saifeldin N Shehata, Maria Deak, Nicholas A Morrice, Eriko Ohta, Roger W Hunter, Vera M Kalscheuer, Kei Sakamoto
ABSTRACT
PCTAIRE-1 [also known as cyclin-dependent kinase 16 (CDK16)] is implicated in various physiological processes such as neurite outgrowth and vesicle trafficking; however, its molecular regulation and downstream targets are largely unknown. Cyclin Y has recently been identified as a key interacting/activating cyclin for PCTAIRE-1; however, the molecular mechanism by which it activates PCTAIRE-1 is undefined. In the present study, we initially performed protein sequence analysis and identified two candidate phosphorylation sites (Ser(12) and Ser(336)) on cyclin Y that might be catalysed by PCTAIRE-1. Although in vitro peptide analysis favoured Ser(12) as the candidate phosphorylation site, immunoblot analysis of cell lysates that had been transfected with wild-type (WT) or kinase-inactive (KI) PCTAIRE-1 together with WT or phospho-deficient mutants of cyclin Y suggested Ser(336), but not Ser(12), as a PCTAIRE-1-dependent phosphorylation site. Monitoring phosphorylation of Ser(336) may provide a useful read-out to assess cellular activity of PCTAIRE-1 in vivo; however, a phospho-deficient S336A mutant displayed normal interaction with PCTAIRE-1. Unbiased mass spectrometry and targeted mutagenesis analysis of cyclin Y identified key phosphorylation sites (Ser(100) and Ser(326)) required for 14-3-3 binding. Recombinant WT cyclin Y, but not a S100A/S326A mutant, prepared in COS-1 cells co-purified with 14-3-3 and was able to activate bacterially expressed recombinant PCTAIRE-1 in cell-free assays. Finally, we observed that recently identified PCTAIRE-1 variants found in patients with intellectual disability were unable to interact with cyclin Y, and were inactive enzymes. Collectively, the present work has revealed a new mechanistic insight into activation of PCTAIRE-1, which is mediated through interaction with the phosphorylated form of cyclin Y in complex with 14-3-3.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Tetraethylammonium borohydride, technical, ≥95% (T)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O