Merck
  • Recombinant HE4 protein promotes proliferation of pancreatic and endometrial cancer cell lines.
Recombinant HE4 protein promotes proliferation of pancreatic and endometrial cancer cell lines.
Oncology reports (2015-10-27)
Qinsheng Lu, Haibin Chen, Christopher Senkowski, Jianhao Wang, Xue Wang, Steven Brower, Wayne Glasgow, David Byck, Shi-Wen Jiang, Jinping Li
ABSTRACT
Pancreatic adenocarcinoma is one of the most deadly malignancies, and endometrial cancer represents the most common gynecologic cancer in the USA. Better understanding on the pathologic mechanisms and pathways is required for effective treatment of these malignancies. Recently, human epididymis protein 4 (HE4 or WFDC2), a secretory glycoprotein, was found to be overexpressed in pancreatic and endometrial cancers. In addition, studies have shown that HE4 overexpression in endometrial cancer cell lines led to faster cancer progression in a mouse subcutaneous model. These findings raise a question on the role(s) of secretory, extracellular HE4 in cancer development. In the present study, we found that treatment of pancreatic and endometrial cancer cell lines with purified, extracellular HE4 protein led to a significant increase in cell viability and proliferation. Moreover, extracellular HE4 protein was able to increase DNA synthesis, and modulate the mRNA and protein levels of cell cycle marker PCNA and cell cycle inhibitor p21. These effects appeared to be robust and sustainable and required a relatively low concentration of HE4 protein. The findings indicated the secreted, extracellular HE4 may carry some physiopathological functions. Via paracrine/endocrine actions, circulatory HE4 produced by malignant cells may contribute to pancreatic and endometrial cancer progression and/or metastasis.
Product Description
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, BioUltra, ≥99%
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, ≥99% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets