Merck
  • Size characterization of commercial micelles and microemulsions by Taylor dispersion analysis.
Size characterization of commercial micelles and microemulsions by Taylor dispersion analysis.
International journal of pharmaceutics (2015-07-05)
Joseph Chamieh, Florian Davanier, Vincent Jannin, Frédéric Demarne, Hervé Cottet
ABSTRACT
In this work, Taylor dispersion analysis was applied to the measurement of micelles (or microdroplets) molecular diffusion coefficient in micellar (or microemulsion) systems based on neutral/anionic/cationic or zwitterionic surfactants. The choice of the micellar marker and the influence the surfactant/marker concentrations on this determination are studied. Experimental results are compared to those derived from the literature using other experimental techniques. Taylor dispersion analysis, experienced in narrow capillaries, was found to be an efficient and suitable method for micelle (or microdroplet) size measurement due to: the low sample consumption, the absence of filtration requirement of the sample, the broad range of size determination (with no lower limit down to angstroms), the simplicity of the protocol, the possibility to measure the viscosity of surfactant solutions in given conditions and the determination of the weight-average micelle hydrodynamic radius. Application to the size-characterization of commercial microemulsions (Gelucire(®) 44/14), used as an excipient in the pharmaceutical formulation, is provided with a comparison to DLS measurements. It was found that the polydispersity in size of the micelle did not influence the Gaussian peak shape of the taylorgram due to rapid surfactant exchange compared to the time-scale of the experiments (a few minutes).
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
1-Pentanol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Amyl alcohol, ≥99%, FG
Sigma-Aldrich
1-Pentanol, ACS reagent, ≥99%
Sigma-Aldrich
Hexaethylene glycol monododecyl ether, BioXtra, ≥98.0% (TLC)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri