Merck
  • Analysis of tandem E-box motifs within human Complement receptor 2 (CR2/CD21) promoter reveals cell specific roles for RP58, E2A, USF and localized chromatin accessibility.
Analysis of tandem E-box motifs within human Complement receptor 2 (CR2/CD21) promoter reveals cell specific roles for RP58, E2A, USF and localized chromatin accessibility.
The international journal of biochemistry & cell biology (2015-03-31)
Mark N Cruickshank, James Dods, Rhonda L Taylor, Mahdad Karimi, Emily J Fenwick, Elizabeth A Quail, Alexander J Rea, V Michael Holers, Lawrence J Abraham, Daniela Ulgiati
ABSTRACT
Complement receptor 2 (CR2/CD21) plays an important role in the generation of normal B cell immune responses. As transcription appears to be the prime mechanism via which surface CR2/CD21 expression is controlled, understanding transcriptional regulation of this gene will have broader implications to B cell biology. Here we report opposing, cell-context specific control of CR2/CD21 promoter activity by tandem E-box elements, spaced 22 bp apart and within 70 bp of the transcription initiation site. We have identified E2A and USF transcription factors as binding to the distal and proximal E-box sites respectively in CR2-positive B-cells, at a site that is hypersensitive to restriction enzyme digestion compared to non-expressing K562 cells. However, additional unidentified proteins have also been found to bind these functionally important elements. By utilizing a proteomics approach we have identified a repressor protein, RP58, binding the distal E-box motif. Co-transfection experiments using RP58 overexpression constructs demonstrated a specific 10-fold repression of CR2/CD21 transcriptional activity mediated through the distal E-box repressor element. Taken together, our results indicate that repression of the CR2/CD21 promoter can occur through one of the E-box motifs via recruitment of RP58 and other factors to bring about a silenced chromatin context within CR2/CD21 non-expressing cells.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Aldrich® Imperial mini-spill kits, capacity 1.8 gal (6.8 L)
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Aldrich® Imperial mini-spill kits, capacity 3.6 gal (13.6 L)
Mini chemical spill kit
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Tributylphosphine, ≥93.5% (Tri-N-butylphosphine, GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Tributylphosphine, mixture of isomers, 97%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
SAFC
BIS-TRIS
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri