Merck
  • Preparation of human tau exon-2- and -10-specific monoclonal antibodies for the recognition of brain tau proteins in various mammals.
Preparation of human tau exon-2- and -10-specific monoclonal antibodies for the recognition of brain tau proteins in various mammals.
International journal of molecular medicine (2015-06-06)
Cao Chen, Yan Lv, Qi Shi, Bao-Yun Zhang, Li-Na Chen, Kang Xiao, Jing Sun, Xiao-Ping Dong
ABSTRACT
The aggregations of tau protein in brain tissue have been described in a large number of neurodegenerative diseases; however, due to the lack of tau isoform- or exon-specific antibodies, the exact situations under which various brain tau isoforms can be found and their exact contributions during disease progression remain unknown. Therefore, in this study, we prepared tau exon-specific monoclonal antibodies (mAbs) that recognize different mammalian tau isoforms. Briefly, 3 Balb/c mice were separately immunized (3 mice per antigen) with the recombinant GST-fusion proteins, GST-tE2 and GST-tE10. Two hybridoma cell lines, 4A8 and 3E12, secreting antibodies against human tau exon-2 and -10 were established using the hybridoma technique. The sensitivity and specificity of the prepared mAbs were evaluated using indirect ELISA and western blot analysis. The ability of the prepared mAbs, 4A8 and 3E12, to recognize endogenous tau protein in the brain tissues of various mammals was estimated by immunoprecipitation. Based on the results of various verification methods, we found that the prepared mAbs, 4A8 and 3E12, not only specifically reacted with the individual recombinant GST tau exon fusion proteins, but also correctly recognized the recombinant human tau isoforms containing respective exon sequences, as shown by western blot analysis. Furthermore, western blot analysis and immunoprecipitation assays verified that the mAbs, 4A8 and 3E12, recognized endogenous tau proteins in human brain tissue, as well as tau proteins in a series of mammalian tissues, including goat, bovine, rabbit, hamster and mouse. Thus, in the present study, using the hybridoma technique, we successfully prepared the mAbs, 4A8 against tau exon-2 and 3E12 against tau exon-10, which provide useful tools for determining potential alternations of tau isoforms in neurodegenerative diseases.
Product Description
Roche
IsoStrip Mouse Monoclonal Antibody Isotyping Kit, sufficient for 10 tests
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)