Merck
  • Myclobutanil worsens nonalcoholic fatty liver disease: An in vitro study of toxicity and apoptosis on HepG2 cells.
Myclobutanil worsens nonalcoholic fatty liver disease: An in vitro study of toxicity and apoptosis on HepG2 cells.
Toxicology letters (2016-10-25)
Antonietta Stellavato, Monica Lamberti, Anna Virginia Adriana Pirozzi, Francesca de Novellis, Chiara Schiraldi
ABSTRACT
Myclobutanil is a conazole class fungicide widely used as an agrichemical. It is approved for use on fruit, vegetables and seed commodities in the EU and elsewhere to control fungi such as Ascomycetes, Fungi Imperfecti and, Basidiomycetes. Its widespread use has raised the issue of possible health risks for agrarian communities and the general population, which can be exposed to residues present in food and drinking water. The toxicities identified include adverse effects on liver and kidney and on the development of male reproductive organs. Since the liver is the first-line organ in the defense against xenobiotics, toxic effects on hepatic metabolism cause degeneration, necrosis, and tissue hypertrophy. Therefore, we investigated myclobutanil's effects on the human liver cell line HepG2. We found that myclobutanil increases the amount of fatty acids in these hepatic cells, as evaluated with Oil Red O staining, and progressively reduces cell viability from 1ppm to 500ppm. Analysis of biomarkers such as Bcl-xL/Bak and Mcl-1/Bak confirmed activation of cell death pathways at low doses. Therefore, myclobutanil may play an important role in the pathogenesis and progression of chronic hepatocellular diseases in humans.
Product Description
Supelco
Myclobutanil, PESTANAL®, analytical standard
Pricing and availability is not currently available.