- Effects of Carbopol
Effects of Carbopol
International journal of nanomedicine (2016-11-24)
Yin Zheng, Wu-Qing Ouyang, Yun-Peng Wei, Shahid Faraz Syed, Chao-Shuang Hao, Bo-Zhen Wang, Yan-Hong Shang
PMID27877042
ABSTRACT
Nanoemulsions (NEs) are used as transdermal drug delivery systems for systematic therapeutic purposes. We hypothesized that the skin permeation profile of an NE could be modulated by incorporating it into a hydrogel containing differing proportions of thickening agent. The objectives of this study were as follows: 1) to determine the stability and skin irritability of NE gels (NGs) containing 1%, 2%, and 3% (w/w) Carbopol