Merck
  • Kruppel-like factor 4 is critical for transcriptional control of cardiac mitochondrial homeostasis.
Kruppel-like factor 4 is critical for transcriptional control of cardiac mitochondrial homeostasis.
The Journal of clinical investigation (2015-08-05)
Xudong Liao, Rongli Zhang, Yuan Lu, Domenick A Prosdocimo, Panjamaporn Sangwung, Lilei Zhang, Guangjin Zhou, Puneet Anand, Ling Lai, Teresa C Leone, Hisashi Fujioka, Fang Ye, Mariana G Rosca, Charles L Hoppel, P Christian Schulze, E Dale Abel, Jonathan S Stamler, Daniel P Kelly, Mukesh K Jain
ABSTRACT
Mitochondrial homeostasis is critical for tissue health, and mitochondrial dysfunction contributes to numerous diseases, including heart failure. Here, we have shown that the transcription factor Kruppel-like factor 4 (KLF4) governs mitochondrial biogenesis, metabolic function, dynamics, and autophagic clearance. Adult mice with cardiac-specific Klf4 deficiency developed cardiac dysfunction with aging or in response to pressure overload that was characterized by reduced myocardial ATP levels, elevated ROS, and marked alterations in mitochondrial shape, size, ultrastructure, and alignment. Evaluation of mitochondria isolated from KLF4-deficient hearts revealed a reduced respiration rate that is likely due to defects in electron transport chain complex I. Further, cardiac-specific, embryonic Klf4 deletion resulted in postnatal premature mortality, impaired mitochondrial biogenesis, and altered mitochondrial maturation. We determined that KLF4 binds to, cooperates with, and is requisite for optimal function of the estrogen-related receptor/PPARγ coactivator 1 (ERR/PGC-1) transcriptional regulatory module on metabolic and mitochondrial targets. Finally, we found that KLF4 regulates autophagy flux through transcriptional regulation of a broad array of autophagy genes in cardiomyocytes. Collectively, these findings identify KLF4 as a nodal transcriptional regulator of mitochondrial homeostasis.
Product Description
Millipore
EZview Red Anti-HA Affinity Gel
Sigma-Aldrich
ANTI-FLAG® M2-Peroxidase (HRP) antibody, Mouse monoclonal, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Anti-HA−Peroxidase antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Millipore
EZview Red ANTI-FLAG® M2 Affinity Gel, clone M2