Merck
  • Semi-quantitative analysis of multiple chemical mixtures in solution at trace level by surface-enhanced Raman Scattering.
Semi-quantitative analysis of multiple chemical mixtures in solution at trace level by surface-enhanced Raman Scattering.
Scientific reports (2017-07-25)
Sumeng Zou, Mengjing Hou, Jianghao Li, Lingwei Ma, Zhengjun Zhang
ABSTRACT
Surface-enhanced Raman scattering (SERS) technology combines with chemometric method of principal component analysis (PCA) was used to calculate the composition of chemical mixtures in solution. We reported here that there exists composition discrepancy between molecules in solution and molecules adsorbed on Ag@Al
Sigma-Aldrich
4-Mercaptopyridine, 95%
Pricing and availability is not currently available.