Merck
  • Optimal Pre-treatment for Acute Exposure to the Organophosphate Dicrotophos.
Optimal Pre-treatment for Acute Exposure to the Organophosphate Dicrotophos.
Current pharmaceutical design (2016-11-02)
Dietrich E Lorke, Syed M Nurulain, Mohamed Y Hasan, Kamil Kuca, Georg A Petroianu
ABSTRACT
Reversible cholinesterase inhibitors, when given prophylactically before exposure to organophosphates, are able to decrease organophosphate-induced mortality. However, the efficacy of pyridostigmine, the only pre-treatment substance approved by the US Federal Drug Administration, is unsatisfactory. In search of a better prophylactic compound, we determined in vivo the protection conferred by five cholinesterase inhibitors (ranitidine, physostigmine, tacrine, K-27 and pyridostigmine), which were administered in equitoxic dosage (1/4 of LD01) 30 minutes before exposure to the organophosphate dicrotophos. Efficacy was measured in rats by Cox analysis calculating the relative risk of death (RR), RR being 1 for the reference group which received dicrotophos and no prophylaxis. K-27 (RR=0.06), physostigmine (RR=0.15), pyridostigmine (RR=0.22) and tacrine (RR=0.28) significantly (p ≤ 0.05) reduced dicrotophos-induced mortality in comparison to the reference group (dicrotophos without pre-treatment), whereas ranitidine (RR=0.86) had no significant influence. The experimental oxime K-27, when given before dicrotophos exposure, conferred the best in vivo protection. This was significantly (p ≤ 0.05) more efficacious than pre-treatment with any other tested compound. The differences in efficacy between the second best compound, physostigmine, and the less efficacious substances (tacrine and pyridostigmine) were also statistically significant. These data indicate that K-27 can be considered a very efficacious prophylactic agent for organophosphate exposure.
Product Description
Supelco
Dicrotophos, PESTANAL®, analytical standard
Pricing and availability is not currently available.