- JQ1 synergizes with the Bcl-2 inhibitor ABT-263 against
JQ1 synergizes with the Bcl-2 inhibitor ABT-263 against
Oncotarget (2017-11-22)
Huogang Wang, Bo Hong, Xuemin Li, Ke Deng, Hong Li, Vivian Wai Yan Lui, Wenchu Lin
PMID29156797
ABSTRACT
Small cell lung cancer (SCLC) is a clinically aggressive cancer with very poor prognosis. Amplification of