Merck
PH005485
2-Chloro-5-(hydrazinocarbonyl)-N,N-dimethylbenzenesulfonamide
AldrichCPR
Wybierz wielkość
Zmień widok
Informacje o tej pozycji
Wzór liniowy:
C9H12ClN3O3S
Numer CAS:
eCl@ss:
32150312
Zaloguj się – aby sprawdzić ceny
Pomoc techniczna
Pozwól nam pomóc
SMILES string
ClC1=C(S(N(C)C)(=O)=O)C=C(C(NN)=O)C=C1
InChI
1S/C9H12ClN3O3S/c1-13(2)17(15,16)8-5-6(9(14)12-11)3-4-7(8)10/h3-5H,11H2,1-2H3,(H,12,14)
Other Notes
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Not applicable
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)