Przejdź do zawartości
R525944
{2-[(E)-({[3-(2-THIENYL)-1H-PYRAZOL-4-YL]CARBONYL}HYDRAZONO)METHYL]PHENOXY}ACETIC ACID
AldrichCPR
Informacje o tej pozycji
Wzór liniowy:
C17H14N4O4S
Numer CAS:
Masa cząsteczkowa:
370.39
MDL number:
Pomoc techniczna
Pozwól nam pomóc
Other Notes
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)