Merck
Z106917
PE bench-top base
size L
Informacje o tej pozycji
NACRES:
NB.35
UNSPSC Code:
41103406
eCl@ss:
32020603
Pomoc techniczna
Pozwól nam pomóc
size
L
size
24 in. × 34 1/2 in.
thickness
1/2 in.
General description
BENCH-TOP BASE is not autoclavable and PP BENCH TRAY and rod are autoclavable.
Rigid. Keeps AtmosBag in place. Fits inside respective bag.
Still not finding the right product?
Explore all of our products under PE bench-top base
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)