Merck
Z558281
Suba-Seal® septa
blue rubber, Suba-Seal® 49, fits neck I.D., 24 mm
Informacje o tej pozycji
UNSPSC Code:
41121800
NACRES:
NB.35
eCl@ss:
32119211
Material:
blue rubber
Pomoc techniczna
Pozwól nam pomóc
material
blue rubber
feature
fits neck I.D. 24 mm
manufacturer/tradename
Suba-Seal® 49
General description
Well-known as the best quality rubber turn-over closure with serrated stopper, Suba-Seal brand is available exclusively from Sigma-Aldrich. Ideal for air and moisture-sensitive chemistry procedures. After piercing with non-coring needle, rubber closes puncture, providing air and moisture tight seal to protect contents of vessel or container from atmosphere.
This closure has turnover flange molded to grip outside of container neck promoting a double seal in conjunction with annular serrations on hollow plug. Annular serrations depress themselves against the inside wall of the bottle neck, making each serration a suctions sealing point.
Storage Note for Natural Rubber Septa: Store septa in a closed bag out of sunlight to retard blooming, which is the formation of a whitish film on the surface of the rubber. Blooming is a natural process and does not affect performance. Remove visible bloom by washing or wiping off before use.
Note: For extra security when using large bore transfer needles, fasten septum to bottle or flask opening with a wrap-it tie. See ties and accessory tool listings.
Legal Information
Suba-Seal is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under Suba-Seal® septa
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)