Informacje o tej pozycji
NACRES:
NA.28
UNSPSC Code:
12352200
eCl@ss:
32160703
Form:
liquid
Storage condition:
OK to freeze
Pomoc techniczna
Pozwól nam pomócform
liquid
Quality Segment
specific activity
≥1 unit/µg protein
storage condition
OK to freeze
storage temp.
−70°C
General description
Recombinant, human ACE ectodomain (amino acids 30-1261) fused at the C-terminus to a HIS TAG® sequence and expressed in NSO cells. ACE is a zinc metallopeptidase that cleaves the C-terminal dipeptide from angiotensin I to produce the potent vasopressor octapeptide angiotensin II. ACE gene polymorphism has been a recent topic in Alzheimer′s disease studies. ACE is a type I membrane protein and exists in two isoforms, somatic and germinal. By SDS-PAGE, the apparent molecular weight of the glycosylated protein appears larger to be than 120 kDa under both reducing and non-reducing conditions.
Legal Information
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?