Merck
186-M
Congo Red
HARLECO® For the detection of amyloid deposits
Synonim(y):
Congo Red, Direct Red
Informacje o tej pozycji
Wzór empiryczny (zapis Hilla):
C32H22N6Na2O6S2
Numer CAS:
Masa cząsteczkowa:
696.66
EC Index Number:
209-358-4
UNSPSC Code:
12352107
MDL number:
Pomoc techniczna
Pozwól nam pomóc
vapor pressure
<1 Pa
Quality Segment
pH
6.7 (20 °C, 10 g/L in H2O)
mp
>360 °C
solubility
25 g/L
storage temp.
10-30°C
InChI
1S/C32H24N6O6S2.2Na/c33-31-25-7-3-1-5-23(25)29(45(39,40)41)17-27(31)37-35-21-13-9-19(10-14-21)20-11-15-22(16-12-20)36-38-28-18-30(46(42,43)44)24-6-2-4-8-26(24)32(28)34;;/h1-18H,33-34H2,(H,39,40,41)(H,42,43,44);;/q;2*+1/p-2
InChI key
IQFVPQOLBLOTPF-UHFFFAOYSA-L
Application
Certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Used for cytoplasmic staining of paraffin sections fixed in Bouin’s fluid. Also a general tissue stain for use as a cytoplasmic counterstain.
HARLECO® For the detection of amyloid deposits
Legal Information
HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under Congo Red
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)