Merck
71726
DNA pIEx/Bac-3 - Novagen
Novagen′s pIEx/Bac vectors are dual-purpose vectors that offer unique flexibility for cloning and expression of proteins in Spodoptera-derived insect cells.
Synonim(y):
wektor, wektor ekspresji
Informacje o tej pozycji
UNSPSC Code:
41106600
NACRES:
NA.54
Pomoc techniczna
Pozwól nam pomóc
Quality Segment
Disclaimer
This product is sold for internal research use only. Any commercial use of this product, its components, and/or any derivatives thereof (including but not limited to proteins produced using the product or its components) (together and hereinafter the “EMD Product”) requires signature of a written commercial use agreement with EMD Millipore Corporation or its successor-in-interest. Commercial use shall include but not be limited to: (1) use of the EMD Product to manufacture products for sale to third parties; (2) use of the EMD Product to provide services, information, or data to third parties in exchange for consideration; (3) use of the EMD Product for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the EMD Product, whether or not such EMD Product is resold for research use. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of the EMD Product. Please direct any questions on these use restrictions to: licensing@milliporesigma.com.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)