Przejdź do zawartości
Merck

AB356487

Anti-pan-fMet

Synonim(y):

N-terminal formyl-methionine, Nt-fMet

Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.

Wybierz wielkość

Zmień widok

Informacje o tej pozycji

UNSPSC Code:
12352203
NACRES:
NA.46
eCl@ss:
32160702
Species reactivity:
E. coli
Technique(s):
ELISA: suitable, western blot: suitable
Application:
ELISA, WB
Citations:
-
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomóc


biological source

rabbit

Quality Segment

antibody form

purified antibody

antibody product type

primary antibodies

mol wt

calculated mol wt 20~50 kDa, observed mol wt ~45 kDa

species reactivity

E. coli

species reactivity (predicted by homology)

all

packaging

antibody small pack of 100

technique(s)

ELISA: suitable, western blot: suitable

isotype

IgGκ

UniProt accession no.

storage temp.

-10 to -25°C

Immunogen

KLH-conjugated, N-terminal formylmethionine synthetic peptide (fMet-Xaa-Cys).

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from DH5 cells treated with Actinonin.

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected N-terminal formylmethionine bearing proteins in lysate from DH5 cells treated with Actinonin but, not in DH5 cells treated with Ethanol.

Tested Applications

Western Blotting Analysis: A representative lot detected fMet (N-terminal formylmethionine) in Western Blotting application.

ELISA: A representative lot of this antibody detected GST-tagged recombinant fMD-D2(3-11) in ELISA application.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

This polyclonal antibody specifically detects fMet (N-terminal formylmethionine) bearing proteins and their derivatives.

Physical form

Rabbit polyclonal antibody in serum without azide.

Preparation Note

Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
N-terminal formyl-methionine (fMet) is a unique amino acid that serves as the initiating residue in protein synthesis in bacteria and mitochondria, playing a pivotal role in various cellular processes. Structurally, fMet is characterized by the addition of a formyl group to the methionine, which influences the stability and function of nascent polypeptides. Nt-fMet is produced pre-translationally by formyltransferases, which use 10-formyltetrahydrofolate as a cosubstrate. By contrast, proteins synthesized by cytosolic ribosomes of eukaryotes are presumed to bear unformylated N-terminal Met (Nt-Met). The unformylated Nt-Met residue of eukaryotic proteins is often co-translationally Nt acetylated, a modification that creates specific degradation signals, Ac/Ndegrons, which are targeted by the Ac/N-end rule pathway. Functionally, fMet acts as a degradation signal in the N-end rule pathway, guiding the proteolytic fate of proteins. In eukaryotic cells, fMet-containing proteins can be synthesized in mitochondria and the cytosol, with recent studies indicating substantial upregulation of fMet-containing proteins in various cancer cell lines, suggesting a potential link to tumor progression and metastasis. Subcellular localization of fMet-containing proteins is crucial, as their presence in mitochondrial membranes is essential for the assembly of respiratory complexes. Dysregulation of fMet metabolism has been implicated in several diseases, including cancer and mitochondrial disorders. Both bacterial and eukaryotic ribosomes can initiate protein synthesis with formylmethionine (fMet), but detecting fMet-bearing peptides and fMet-bearing proteins has been challenging due to the lack of effective anti-fMet antibodies. This anti-pan-fMet polyclonal Ab specifically detects N-terminal formylmethionine-bearing proteins and their derivatives. It is suitable for western blotting, dot-blotting and ELISA validations. (Ref.: Kim, D., et al. (2025). BioTechniques. 77(2); 46-55; Kim, D., et al. (2023). J. Bio. Chem. 299(5);104652; Kim, JM., et al. (2018). Science. 362(6418); eaat0174).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Ta strona może zawierać tekst przetłumaczony maszynowo.


Still not finding the right product?

Wypróbuj nasze narzędzie Narzędzie selektora produktów, aby zawęzić opcje.


Temperatura zapłonu (°F)

Not applicable

Temperatura zapłonu (°C)

Not applicable

Klasa składowania

10 - Combustible liquids



Wybierz jedną z najnowszych wersji:

Certyfikaty analizy (CoA)

Lot/Batch Number

Nie widzisz odpowiedniej wersji?

Jeśli potrzebujesz konkretnej wersji, możesz wyszukać konkretny certyfikat według numeru partii lub serii.

Masz już ten produkt?

Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.

Odwiedź Bibliotekę dokumentów