Merck
IM51
Anty-MMP-2 (Ab-4) mysie mAb (75-7F7)
Informacje o tej pozycji
NACRES:
NA.41
UNSPSC Code:
51111800
Species reactivity:
(bovine, human, mouse, not rat)
Species reactivity:
-
Application:
—
Citations:
-
Pomoc techniczna
Pozwól nam pomóc
biological source
mouse
Quality Segment
form
liquid
species reactivity
(bovine, human, mouse, not rat)
storage condition
OK to freeze
dilution
(Frozen Sections
Immunoblotting (1 µg/mL,
Paraffin Sections )
isotype
IgG1
storage temp.
−20°C
General description
Purified mouse monoclonal antibody. Recognizes the ~72 kDa latent and the ~66 kDa active forms of MMP-2.
Immunogen
Human
Application
Frozen Sections Immunoblotting (1 µg/mL, Paraffin Sections
Klasa składowania
10 - Combustible liquids
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)