Przejdź do zawartości
Merck

MABS2483

Anti-Dectin-2 Antibody, clone 17A1D10

Synonim(y):

C-type lectin domain family 6 member A, C-type lectin superfamily member 10, DECTIN2, Dectin-2, Dendritic cell-associated C-type lectin 2, DC-associated C-type lectin 2

Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.

Wybierz wielkość

Zmień widok

Informacje o tej pozycji

UNSPSC Code:
12352203
NACRES:
NA.46
eCl@ss:
32160702
Clone:
17A1D10, monoclonal
Species reactivity:
monkey, human
Application:
ELISA, FACS, inhibition assay, surface plasmon resonance (SPR)
Citations:
-
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomóc


biological source

mouse

Quality Segment

antibody form

purified antibody

antibody product type

primary antibodies

clone

17A1D10, monoclonal

mol wt

calculated mol wt 27.51 kDa, observed mol wt ~N/A kDa

purified by

using protein G

species reactivity

monkey, human

packaging

antibody small pack of 100

technique(s)

ELISA: suitable, flow cytometry: suitable, inhibition assay: suitable, surface plasmon resonance (SPR): suitable

isotype

IgG2aκ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

storage temp.

-10 to -25°C

Gene Information

human ... CLEC6A(50856)

General description

C-type lectin domain family 6 member A (UniProt: Q6EIG7; also known as C-type lectin superfamily member 10, Dendritic cell-associated C-type lectin 2, DC-associated C-type lectin 2, Dectin-2) is encoded by the Clec6a (also known as CLECSF10, DECTIN2) gene (Gene ID: ) in human. Dectin-2 plays a critical role in the innate and adaptive immune system by recognizing pathogen-associated molecular patterns (PAMPs) and initiating immune responses. This receptor is an ITAM-coupled C-type lectin receptor (CLR); a single-pass type II transmembrane protein. It features a carbohydrate recognition domain (CRD) that allows it to bind specifically to high-mannose polysaccharides found on the surface of various pathogens, including fungi and some bacteria. It is characterized by a single extracellular (aa 42-209) C-type lectin-like domain (CTLD) connected by a transmembrane region (aa 22-41) to a short cytoplasmic tail (aa 1-20) that lacks a clear intracellular signaling motif. Association of Dectin-2 with adaptor proteins, such as the Fc receptor (FcR ) chain or DNAX-activation protein 12 (DAP12), is required for the engagement to Syk pathway, leading to the activation of NF- B and the production of pro-inflammatory cytokines. Its expression is particularly high in the spleen and lymph nodes and primarily expressed on dendritic cells and macrophages. Dectin-2 has been implicated in various diseases, particularly those involving fungal infections due to its role in recognizing fungal cell wall components. dysregulation of Dectin-2 signaling has been associated with inflammatory and autoimmune diseases, highlighting its importance in maintaining immune homeostasis. Dectin-2 plays critical roles in the pathogenesis of allergic diseases. (Ref.: Malamud, M., and Brown, G. D., (2024) EMBO Rep, 25, 5239-5264); Chen, M.H., et al., (2019) J. Biomed. Sci. 26:97; Angelina, A., et al., (2023) Curr Allergy and Asthma Rep. 23, 141-151)

Immunogen

hFc-tagged, full-length human Dectin-2.

Application

Quality Control Testing

Evaluated by Enzyme Immunoassay (ELISA) with hFc-tagged, full-length, recombinant, human Dectin-2.

Enzyme Immunoassay (ELISA): A representative lot of this antibody detected hFc-tagged, full-length, recombinant, human Dectin-2.

Tested Applications

Surface plasmon resonance: Chen, M-H., et al. (2019). J Biomed Sci. 26(1):97.

ELISA Analysis: A representative lot detected Dectin-2 in ELISA application (Chen, M-H., et al. (2019). J Biomed Sci. 26(1):97).

Inhibition Assay: A representative lot of this antibody inhibited house dust mite binding to Dectin-2. (Chen, M-H., et al. (2019). J Biomed Sci. 26(1):97).

Flow Cytometry Analysis: A representative lot detected Dectin-2 in Flow Cytometry application (Chen, M-H., et al. (2019). J Biomed Sci. 26(1):97).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone 17A1D10 is a mouse monoclonal antibody that detects Dectin-2.

Physical form

Purified mouse monoclonal antibody IgG2a in PBS without preservatives.

Preparation Note

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Ta strona może zawierać tekst przetłumaczony maszynowo.


Still not finding the right product?

Wypróbuj nasze narzędzie Narzędzie selektora produktów, aby zawęzić opcje.


Klasa składowania

12 - Non Combustible Liquids

WGK

WGK 3

Temperatura zapłonu (°F)

Not applicable

Temperatura zapłonu (°C)

Not applicable



Wybierz jedną z najnowszych wersji:

Certyfikaty analizy (CoA)

Lot/Batch Number

Nie widzisz odpowiedniej wersji?

Jeśli potrzebujesz konkretnej wersji, możesz wyszukać konkretny certyfikat według numeru partii lub serii.

Masz już ten produkt?

Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.

Odwiedź Bibliotekę dokumentów