Merck
OP67
Mysie mAb anty-c-ErbB2/c-Neu (Ab-6) (OD-3)
Informacje o tej pozycji
NACRES:
NA.41
UNSPSC Code:
51111800
Species reactivity:
(human, not mouse, not rat)
Species reactivity:
-
Application:
—
Citations:
-
Pomoc techniczna
Pozwól nam pomóc
biological source
mouse
Quality Segment
form
liquid
species reactivity
(human, not mouse, not rat)
storage condition
do not freeze
dilution
(Immunoblotting (10 µg/mL)
Frozen Sections (not recommended)
Paraffin Sections (not recommended)
Immunofluorescence (not recommended)
Immunoprecipitation (not recommended))
isotype
IgM
storage temp.
2-8°C
General description
Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing with SP2/0 mouse myeloma cells. Recognizes the ~185 kDa full-length and the ~97-115 kDa truncated forms of c-ErbB2/c-Neu.
Immunogen
Human
Application
Immunoblotting (10 µg/mL)Frozen Sections (not recommended)Paraffin Sections (not recommended)Immunofluorescence (not recommended)Immunoprecipitation (not recommended)
Klasa składowania
12 - Non Combustible Liquids
Not applicable
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)