Informacje o tej pozycji
NACRES:
NA.45
UNSPSC Code:
51111800
Species reactivity:
rabbit
Species reactivity:
-
Application:
—
Citations:
-
Pomoc techniczna
Pozwól nam pomócQuality Segment
biological source
goat
form
liquid
species reactivity
rabbit
storage condition
OK to freeze
dilution
(ELISA (1 µg/mL)
Flow Cytometry (1 µg/mL)
Immunoblotting (0.2 µg/mL)
Immunofluorescence (1 µg/mL)
Immunohistochemistry (1 µg/mL)
Immunoprecipitation (1 µg/sample))
isotype
IgG
storage temp.
−20°C
General description
Immunoaffinity purified goat polyclonal antibody adsorbed against human and bovine serum proteins and conjugated to biotin. Recognizes rabbit IgG.
Immunogen
Rabbit
Application
ELISA (1 µg/mL)Flow Cytometry (1 µg/mL)Immunoblotting (0.2 µg/mL)Immunofluorescence (1 µg/mL)Immunohistochemistry (1 µg/mL)Immunoprecipitation (1 µg/sample)
Klasa składowania
12 - Non Combustible Liquids
Not applicable
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?