Merck
SI1M024A5
MPHL2GCL3 3/PK
Synonim(y):
MPHL2GCL3 3/PK
Informacje o tej pozycji
UNSPSC Code:
23151806
Pomoc techniczna
Pozwól nam pomóc
Nazwa produktu
Millipak® 200 Gamma Gold CapsuleMedia: Sterile Durapore® 0.45 µm, Chemistry: Hydrophilic Polyvinylidene Fluoride (PVDF)Inlet: 1-1/2 in. Sanitary FlangeOutlet: 1-1/2 in. Sanitary Flange
Quality Segment
feature
hydrophilic
manufacturer/tradename
Durapore®
filtration area
1000.0 cm2
impurities
<0.5 EU/mL USP bacterial endotoxins (LAL test, sample aqueous extraction)
matrix
Durapore®
bubble point
≥1790 mbar (26 psig), air with water at 23 °C
fitting
38 mm (1 1/2 in.) inlet sanitary flange, 38 mm (1 1/2 in.) outlet sanitary flange
Legal Information
Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIPAK is a registered trademark of Merck KGaA, Darmstadt, Germany
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)