Merck
CL53
CCS Viscosity Standard
ASTM D 5293 and SAE Specification J325
Synonim(y):
Cold-Cranking Simulator Viscosity Standard
Informacje o tej pozycji
NACRES:
NA.24
UNSPSC Code:
41116107
Pomoc techniczna
Pozwól nam pomóc
grade
certified reference material
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Paragon Scientific Ltd
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
format
mixture
General description
Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon CCS Viscosity Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.
Still not finding the right product?
Explore all of our products under CCS Viscosity Standard
Klasa składowania
10 - Combustible liquids
Co się dzieje?
WGK 1
Temperatura zapłonu (°F)
426.2 - 494.6 °F
Temperatura zapłonu (°C)
219 - 257 °C
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)