Merck
A8129
AICAR
Synonim(y):
5-Aminoimidazole-4-carboxamide 1-β-D-ribofuranoside, Acadesine, N1-(β-D-Ribofuranosyl)-5-aminoimidazole-4-carboxamide
Informacje o tej pozycji
Wzór empiryczny (zapis Hilla):
C9H14N4O5
Numer CAS:
Masa cząsteczkowa:
258.23
EC Number:
220-097-5
UNSPSC Code:
12352204
MDL number:
Pomoc techniczna
Pozwól nam pomóc
SMILES string
NC(=O)c1ncn([C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)c1N
InChI
1S/C9H14N4O5/c10-7-4(8(11)17)12-2-13(7)9-6(16)5(15)3(1-14)18-9/h2-3,5-6,9,14-16H,1,10H2,(H2,11,17)/t3-,5-,6-,9-/m1/s1
InChI key
RTRQQBHATOEIAF-UUOKFMHZSA-N
Biochem/physiol Actions
AICAR is a cell permeable activator of AMP-activated protein kinase (AMPK), a metabolic master regulator that is activated in times of reduced energy availability (high cellular AMP:ATP ratios) and serves to inhibit anabolic processes. In vivo, pharmacologic activation of AMPK with AICAR mimics exercise and triggers insulin-independent glucose uptake by skeletal muscle.
AICAR is a cell permeable activator of AMP-activated protein kinase (AMPK).
Still not finding the right product?
Explore all of our products under AICAR
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)