Przejdź do zawartości
Merck

AMAB92010

Monoclonal Anti-PVALB antibody produced in mouse

Prestige Antibodies® Powered by Atlas Antibodies, clone CL15070, purified antibody (Protein A)

enhanced validation

Ulepszona walidacja

Synonim(y):

D22S749

Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.

Wybierz wielkość

Zmień widok

Informacje o tej pozycji

UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
CL15070, monoclonal
Application:
IHC, WB
Citations:
-
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomóc


Quality Segment

biological source

mouse

conjugate

unconjugated

antibody form

purified antibody (Protein A)

antibody product type

primary antibodies

clone

CL15070, monoclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

species reactivity

mouse

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

concentration

1 mg/mL

technique(s)

immunohistochemistry: 1:2500-1:5000, western blot: 1 μg/mL

pH range

7.2-7.4

isotype

IgG1

Ensembl | human accession no.

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

-10 to -25°C

target post-translational modification

unmodified

Gene Information

human ... PVALB(5816)

General description

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Immunogen

Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence

Application

All Prestige Antibodies Powered by Atlas Antibodies are developedand validated by the HumanProteinAtlas (HPA) project and as a result, are supported by the mostextensive characterization in the industry.


The Human Protein Atlas project can be subdivided into three efforts: Human TissueAtlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generatedin support of the Tissue and Cancer Atlas projects have been tested byimmunohistochemistry against hundreds of normal and disease tissues and throughthe recent efforts of the Human Cell Atlas project, many have been characterizedby immunofluorescence to map the human proteome not only at the tissue levelbut now at the subcellular level. These images and the collection of this vastdata set can be viewed on the Human Protein Atlas (HPA) site by clicking on theImage Gallery link. We also provide Prestige Antibodies® protocolsand other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Physical form

100UL pack size is provided in a solution, in PBS(phosphate-buffered saline), pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Large pack size of 1ml (at 1mg/ml) is available in PBS only, purified, biocide free, carrier free and glycerol free. Order here

Preparation Note

Upon arrival, mix gently and aliquot to avoid repeated freeze-thaw cycles

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany
Ta strona może zawierać tekst przetłumaczony maszynowo.


Still not finding the right product?

— lub —

Wypróbuj nasze narzędzie Narzędzie selektora produktów, aby zawęzić opcje.


Klasa składowania

10 - Combustible liquids



Wybierz jedną z najnowszych wersji:

Certyfikaty analizy (CoA)

Lot/Batch Number

Nie widzisz odpowiedniej wersji?

Jeśli potrzebujesz konkretnej wersji, możesz wyszukać konkretny certyfikat według numeru partii lub serii.

Masz już ten produkt?

Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.

Odwiedź Bibliotekę dokumentów