Informacje o tej pozycji
UNSPSC Code:
41121800
eCl@ss:
32160701
Material:
polystyrene plate, white polystyrene wells (collagen coated)
Size:
1536 wells
Pomoc techniczna
Pozwól nam pomócmaterial
polystyrene plate, white polystyrene wells (collagen coated)
sterility
non-sterile
feature
lid: no
packaging
pack of 10 ea, case of 50 ea
manufacturer/tradename
Corning 3549
technique(s)
cell culture: suitable
size
1536 wells
well volume
up to 8 μL
suitability
suitable for (fluorescent; luminescent assays)
binding type
Collagen I coated surface
General description
Corning 1536 Well Plates
- 1536 well polystyrene plates
- Low background fluorescence, minimal light scatter and reduced crosstalk
- Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
- Opaque walls to prevent well-to-well crosstalk
- Flat well bottoms possess superior flatness -minimizes variability within a plate and between plates
- Round well bottoms for reduced air entrapment and improved CV′s and Z factor
- Raised well bottoms for high sensitivity
- Flood reservior on four sides to reduce instrument contamination
- Suitable for fluorescent and luminescent assays
Legal Information
Corning is a registered trademark of Corning, Inc.
Still not finding the right product?
Explore all of our products under Corning® 1536 well plates
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?