Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
purple , white
feature
8-channel, adjustable
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS120216
volume measuring range
30-300 μL
General description
Features and Benefits
- Internal metal parts for increased accuracy and calibration stability, and long life
- Ergonomic Slight downward slant to tip ejector button reduces hand fatigue, incidence of repetitive strain injury (RSI)
- Easy volume adjustment with locking tab to prevent accidental change
- Separate tip ejector
- Large digit volume display
- Color coded and labeled with volume range
- Both top and side have colored coded components for quick visual confirmation
- CE certified and Manufactured in accordance with ISO08655 and GLP regulations
- Autoclavable
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description adjustable, volume 30-300 μL, purple, 8-channel | description adjustable, volume 5-50 μL, orange, 8-channel | description adjustable, volume 1000-10000 μL, red, single-channel | description adjustable, volume 30-300 μL, purple, single-channel |
| material white , purple | material white , orange | material white , red | material white , purple |
| volume measuring range 30-300 μL | volume measuring range 5-50 μL | volume measuring range 1000-10000 μL | volume measuring range 30-300 μL |
| feature 8-channel, adjustable | feature 8-channel, adjustable | feature single-channel, adjustable | feature single-channel, adjustable |
| manufacturer/tradename Heathrow Scientific HS120216 | manufacturer/tradename Heathrow Scientific HS120215 | manufacturer/tradename Heathrow Scientific HS120106 | manufacturer/tradename Heathrow Scientific HS120103 |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
Still not finding the right product?
Explore all of our products under Pearl™ Pipette
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?