Merck
HS120217
Pearl™ Pipette
adjustable, volume 5-50 μL, orange, 12-channel
Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121500
eCl@ss:
32039090
Material:
orange , white
Pomoc techniczna
Pozwól nam pomóc
material
orange , white
feature
12-channel, adjustable
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS120217
volume measuring range
5-50 μL
General description
The Pearl is a single-channel pipette offering adjustable volume and precision liquid handling. The sleek, ergonomic design and reduced plunger pressure help reduce the chance of Repetitive Stress Injury (RSI), as the pipette requires 2/3 the operation pressure of most standard pipettes. The Pearl is tested in accordance with standards and a calibration certificate is supplied. Manufactured in accordance with ISO08655 and GLP regulations.
Features and Benefits
  • Internal metal parts for increased accuracy and calibration stability, and long life
  • Ergonomic Slight downward slant to tip ejector button reduces hand fatigue, incidence of repetitive strain injury (RSI)
  • Easy volume adjustment with locking tab to prevent accidental change
  • Separate tip ejector
  • Large digit volume display
  • Color coded and labeled with volume range
  • Both top and side have colored coded components for quick visual confirmation
  • CE certified and Manufactured in accordance with ISO08655 and GLP regulations
  • Autoclavable
Legal Information
Pearl is a trademark of Specac, Inc.
Still not finding the right product?
Explore all of our products under Pearl™ Pipette
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)