Informacje o tej pozycji
NACRES:
NB.12
UNSPSC Code:
41122804
eCl@ss:
32032201
Manufacturer/tradename:
Heathrow Scientific HS24323E
Feature:
Holds 120 x 10 mL tubes
Material:
polypropylene
Zaloguj się – aby sprawdzić ceny
Pomoc techniczna
Pozwól nam pomócmaterial
polypropylene
feature
Holds 120 x 10 mL tubes
manufacturer/tradename
Heathrow Scientific HS24323E
size
337x235x91 mm
color
white
General description
Mega Rack High-capacity storage rack for sample and blood collection tubes.
- Fully-assembled, polypropylene Mega Tube Racks are great for high throughput labs requiring large sample storage.
- Sturdy and light weight material for easy transport
- Alphanumeric identification for quick location and retrieval of tubes
- Place different test groupings into one rack and eliminate need for multiple tube racks
- Autoclavable
Legal Information
Mega Rack is a registered trademark of Heathrow Scientific
Still not finding the right product?
Explore all of our products under Mega Rack® tube rack
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?