Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
NACRES:
NB.35
UNSPSC Code:
41123302
Manufacturer/tradename:
Heathrow Scientific HS9913
Feature:
Holds 20 x slides
Material:
cardboard
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomócmaterial
cardboard
feature
Holds 20 x slides
manufacturer/tradename
Heathrow Scientific HS9913
size
208x341x9 mm
color
blue, trim
Features and Benefits
- Easily organize slides with the index on the front cover that corresponds with the number grid inside
- Recessed slide compartments and inside covers protect specimens. Slides cannot be touched when the cover is closed, reducing contamination
- Front protection flaps are attached with durable tape for repeated use. Fold flaps flat under the slide holder to save space when in use
- Thumb cut in slide compartments enables easy removal of slides with slight pressure at one end
- Compartment holder is made from a double layer of densely pressed cardboard. Cover is a single layer cardboard
- Disposable
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description Holds 20 x slides, blue, trim | description Holds 20 x slides, green, trim | description Holds 20 x slides 76x26 mm, black (trim) | description Holds 30 x slides, black, trim |
| feature Holds 20 x slides | feature Holds 20 x slides | feature Holds 20 x slides 76x26 mm | feature Holds 30 x slides |
| manufacturer/tradename Heathrow Scientific HS9913 | manufacturer/tradename Heathrow Scientific HS9914 | manufacturer/tradename Heathrow Scientific HS9902 | manufacturer/tradename Heathrow Scientific HS9930 |
| material cardboard | material cardboard | material black (trim) | material cardboard |
| size 208x341x9 mm | size 208x341x9 mm | size - | size 281x328x10 mm |
| color blue, trim | color green, trim | color - | color black, trim |
Still not finding the right product?
Explore all of our products under Cardboard Slide Tray
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?