Merck
L5664
Laser Tough-Spots®
diam. 3/8 in., orange
Synonim(y):
solvent resistant labels, temperature resistant labels, Diversified Biotech SPOT-1300, coloured label, product identification label
Informacje o tej pozycji
NACRES:
NB.35
UNSPSC Code:
55121611
Manufacturer/tradename:
Diversified Biotech SPOT-1300
Material:
orange , polyolefin
Diam.:
3/8 in.
Packaging:
pack of 3840 ea (192 labels per sheet)
Pomoc techniczna
Pozwól nam pomóc
material
orange , polyolefin
packaging
pack of 3840 ea (192 labels per sheet)
manufacturer/tradename
Diversified Biotech SPOT-1300
diam.
3/8 in.
General description
Tough-Spots are chemically inert, polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths, conventional and cryogenic freezer temperatures, most organic solvents, caustic agents and other challenges without peeling, becoming brittle or degrading. Labels can be printed directly from the laser printer. 20 sheets per pack.
Application
cryogenics
Features and Benefits
  • Chemically inert
  • For storage in temperatures from -196°C to 150°C
  • withstands most organic solvents and caustic agents
Legal Information
Tough-Spots is a registered trademark of Diversified Biotech
Still not finding the right product?
Explore all of our products under Laser Tough-Spots®
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)