Merck
SML3267
Nardosinone
≥98% (HPLC)
Synonim(y):
(3aR,9R,9aR,9bS)-1,3a,4,7,8,9,9a,9b-Octahydro-1,1,9,9a-tetramethyl-5H-naphtho[2,1-c][1,2]dioxol-5-one, Nardosinon
Informacje o tej pozycji
Wzór empiryczny (zapis Hilla):
C15H22O3
Numer CAS:
Masa cząsteczkowa:
250.33
UNSPSC Code:
12352200
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Pomoc techniczna
Pozwól nam pomóc
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
SMILES string
O1OC([C@@H]2[C@H]1CC(=O)C3=CCC[C@H]([C@@]32C)C)(C)C
InChI
1S/C15H22O3/c1-9-6-5-7-10-11(16)8-12-13(15(9,10)4)14(2,3)18-17-12/h7,9,12-13H,5-6,8H2,1-4H3/t9-,12-,13+,15+/m1/s1
InChI key
KXGHHSIMRWPVQM-JWFUOXDNSA-N
Biochem/physiol Actions
Main active ingredient from medical plant Nardostachys jatamansi; anti-inflammatory and anti-hypertrophic in cardiomyocytes
Nardosinone is a main active ingredient from medical plant Nardostachys jatamansi. Nardosinone exhibits varies pharmacological effects including anti-inflammatory and anti-hypertrophic in cardiomyocytes. Nardosinone enhances activity of the nerve growth factor and promotes neural stem cells proliferation and differentiation. The molecular mechanism of its activity remains unclear.
Still not finding the right product?
Explore all of our products under Nardosinone
Not applicable
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)