Merck
SML4388
WSP-1
≥98% (HPLC)
Synonim(y):
6′-Methoxy-3-oxospiro[isobenzofuran-1(3H),9′-[9H]xanthen]-3′-yl 2-(2-pyridinyldithio)benzoate, 3′-Methoxy-3-oxo-3H-spiro[isobenzofuran-1,9′-xanthen]-6′-yl 2-(pyridin-2-yldisulfaneyl)benzoate, H2S Sensor WSP-1, Washington State Probe 1, WSP 1, WSP1
Informacje o tej pozycji
Wzór empiryczny (zapis Hilla):
C33H21NO6S2
Numer CAS:
Masa cząsteczkowa:
591.65
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
protect from light
Pomoc techniczna
Pozwól nam pomóc
Quality Segment
assay
≥98% (HPLC)
form
powder
storage condition
protect from light
color
white to beige
solubility
DMSO: 2 mg/mL, clear (warmed)
storage temp.
2-8°C
Biochem/physiol Actions
WSP-1 is a highly selective turn-on fluorescent sensor for hydrogen sulfide detection in living cells and plant systems, employing a nucleophilic substitution-cyclization mechanism to rapidly sense both exogenous and endogenous H2S. With λex/λem of 476/516 nm and a quantum yield (ϕ) of 0.003, WSP-1 exhibits a remarkable 130-fold fluorescence increase upon H2S binding, achieving a detection limit of 60 nM and demonstrating excellent selectivity over other biothiols like cysteine and glutathione. WSP1 maintains optimal performance across physiological pH ranges (6-9) and stability even at higher pH values, outperforming its counterpart WSP-5 in highly alkaline conditions. Despite its poor water solubility necessitating the use of organic co-solvents or surfactants, WSP-1′s fast response time, low nanomolar sensitivity, and significant fluorescence enhancement make it a valuable tool for real-time H2S imaging and quantification in complex cellular environments, enabling the study of H2S-mediated signaling pathways and potential therapeutic applications in cardiovascular and neurodegenerative diseases.
Not applicable
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)