Merck
Z741925
Cameo™ Syringe Filter
Nylon membrane, pore size 0.1 μm, diam. 30 mm
Informacje o tej pozycji
NACRES:
NB.24
UNSPSC Code:
41104900
Pomoc techniczna
Pozwól nam pomóc
material
Nylon membrane, polypropylene housing
sterility
non-sterile
packaging
pkg of 200 ea
manufacturer/tradename
GVS Life Sciences 1224101
diam.
30 mm
pore size
0.1 μm
General description
Designed to speed up and increase sample volume throughput while reducing back pressure, the hold-up volume is slightly lower due to improved flow channel design and reduced spacing between the supports. The design improvements allow an increase in operating pressure the syringe filters can withstand.
Applications:
Analysis of Biological and Organic Fluids, Analytical Sample Prep, HPLC, Composite assays, Content uniformity, Dissolution testing, Environmental samples, Food Analysis
Legal Information
Cameo is a trademark of GVS S.p.A.
Still not finding the right product?
Explore all of our products under Cameo™ Syringe Filter
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)