Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
100 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 245,00 zł |
Informacje o tej pozycji
245,00 zł
packaging
pkg of 100 ea
technique(s)
gas chromatography (GC): suitable
fitting
female cap for 1/4 in. O.D. column (slip-on)
compatibility
for use with 1/4 in. O.D. column
General description
Application
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description for use with 1/4 in. O.D. column | description for use with 1/8 in. O.D. columns | description for use with 1/16 in. O.D. column | description - |
| compatibility for use with 1/4 in. O.D. column | compatibility for use with 1/8 in. O.D. columns | compatibility for use with 1/16 in. O.D. column | compatibility - |
| packaging pkg of 100 ea | packaging pkg of 100 ea | packaging pkg of 100 ea | packaging - |
| technique(s) gas chromatography (GC): suitable | technique(s) gas chromatography (GC): suitable using :Suitable | technique(s) gas chromatography (GC): suitable | technique(s) - |
| fitting female cap for 1/4 in. O.D. column (slip-on) | fitting female cap for 1/8 in. O.D. column (slip-on) | fitting female cap for 1/16 in. O.D. column (slip-on) | fitting - |
Still not finding the right product?
Explore all of our products under Plastic Column Cap
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?