Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 3260,00 zł |
Informacje o tej pozycji
3260,00 zł
General description
Fiber Holder for Manual Sampling
An adjustable depth guide positions the fiber for sampling and for correct placement in the heated zone of the GC injection port. The fiber can be locked in the exposed position.
Fiber Holder for Automated Sampling or HPLC Analysis
Use this fiber holder with a Varian 8100/8200 AutoSampler or with our SPME/HPLC interface. An SPME upgrade kit is necessary for operation with the Varian AutoSampler - contact Varian Instrument Division for information concerning system requirements.
Fiber Holder for CTC CombiPAL/CombiPAL-xt and Gerstel MPS2
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description for use with Non-CTC CombiPAL™ Type Autosamplers, pk of 1 | description for use with CTC CombiPAL, Gerstel MPS2 Autosamplers, pk of 1 | description for use with Manual Sampling, pk of 1 | description Uncoated, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, orange hub |
| greener alternative category | greener alternative category | greener alternative category | greener alternative category |
| compatibility for use with Non-CTC CombiPAL™ Type Autosamplers | compatibility for use with CTC CombiPAL, Gerstel MPS2 Autosamplers | compatibility for use with Manual Sampling | compatibility Manual Holder |
| technique(s) solid phase microextraction (SPME): suitable | technique(s) solid phase microextraction (SPME): suitable | technique(s) solid phase microextraction (SPME): suitable | technique(s) solid phase microextraction (SPME): suitable |
| sustainability Sustainability features | sustainability Sustainability features | sustainability Sustainability features | sustainability Sustainability features |
| packaging pk of 1 | packaging pk of 1 | packaging pk of 1 | packaging pk of 3 |
| application(s) food and beverages | application(s) food and beverages | application(s) - | application(s) food and beverages |
Still not finding the right product?
Explore all of our products under SPME Fiber Holder
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?