Przejdź do
Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.
Informacje o tej pozycji
Wzór liniowy:
[CH3(CH2)16COO]2Ca
Numer CAS:
Masa cząsteczkowa:
607.02
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
3919698
eCl@ss:
25140904
grade
pharmaceutical primary standard
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
CCCCCCCCCCCCCCCCCC(=O)O[Ca]OC(=O)CCCCCCCCCCCCCCCCC
InChI
1S/2C18H36O2.Ca/c2*1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h2*2-17H2,1H3,(H,19,20);/q;;+2/p-2
InChI key
CJZGTCYPCWQAJB-UHFFFAOYSA-L
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
Ta strona może zawierać tekst przetłumaczony maszynowo.
Still not finding the right product?
Explore all of our products under Calcium stearate
Klasa składowania
11 - Combustible Solids
Co się dzieje?
nwg
Temperatura zapłonu (°F)
Not applicable
Temperatura zapłonu (°C)
Not applicable
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.