Purity
96%
Storage temp.
-20C
Country of Origin
US
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
96%
-20C
US
Choose from one of the most recent versions:
Find documentation for the products that you have recently purchased in the Document Library.