Skip to Content
MilliporeSigma

AABH97CD72AE

AA BLOCKS, INC.

Manool

Synonym(s):

(3R)-5-[(1S,4aS,8aS)-5,5,8a-trimethyl-2-methylenedecahydro-1-naphthalenyl]-3-methyl-1-penten-3-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C20H34O1
CAS Number:
Molecular Weight:
290.49
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C20H34O/c1-7-19(5,21)14-11-16-15(2)9-10-17-18(3,4)12-8-13-20(16,17)6/h7,16-17,21H,1-2,8-14H2,3-6H3/t16-,17-,19-,20+/m0/s1

Inchi Key

CECREIRZLPLYDM-QGZVKYPTSA-N

Purity

95%

Storage temp.

N/A

Country of Origin

US



Still not finding the right product?

Explore all of our products under Manool




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library