Skip to Content
MilliporeSigma

AABH97CD7D65

AA BLOCKS, INC.

21-Deoxycortisol

Synonym(s):

(11beta)-11,17-dihydroxypregn-4-ene-3,20-dione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C21H30O4
CAS Number:
Molecular Weight:
346.47
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C21H30O4/c1-12(22)21(25)9-7-16-15-5-4-13-10-14(23)6-8-19(13,2)18(15)17(24)11-20(16,21)3/h10,15-18,24-25H,4-9,11H2,1-3H3/t15-,16-,17-,18+,19-,20-,21-/m0/s1

Inchi Key

LCZBQMKVFQNSJR-UJPCIWJBSA-N

Purity

95%

Storage temp.

N/A

Country of Origin

US



Still not finding the right product?

Explore all of our products under 21-Deoxycortisol




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library