Skip to Content
MilliporeSigma

AABH97CD7E71

AA BLOCKS, INC.

Ofurace

Synonym(s):

2-chloro-N-(2,6-dimethylphenyl)-N-(2-oxotetrahydro-3-furanyl)acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C14H16O3N1Cl1
CAS Number:
Molecular Weight:
281.74
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C14H16ClNO3/c1-9-4-3-5-10(2)13(9)16(12(17)8-15)11-6-7-19-14(11)18/h3-5,11H,6-8H2,1-2H3

Inchi Key

OWDLFBLNMPCXSD-UHFFFAOYSA-N

Storage temp.

N/A

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library