Skip to Content
MilliporeSigma

AABH97D2BF95

AA BLOCKS, INC.

3-Chloro-2,5-furandione

Synonym(s):

3-chloro-2,5-furandione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H1O3Cl1
CAS Number:
Molecular Weight:
132.5
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Purity

95%

Storage temp.

2~8C

Inchi Code

1S/C4HClO3/c5-2-1-3(6)8-4(2)7/h1H

Inchi Key

CXJAFLQWMOMYOW-UHFFFAOYSA-N

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library