Skip to Content
MilliporeSigma

AABH97D3CD1B

AA BLOCKS, INC.

2-Ethylpiperidine-2-carboxylic acid

Synonym(s):

2-ethyl-2-piperidinecarboxylic acid

Select a Size

Change View

About This Item

Linear Formula:
C8H15O2N1
CAS Number:
Molecular Weight:
157.21
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H15NO2/c1-2-8(7(10)11)5-3-4-6-9-8/h9H,2-6H2,1H3,(H,10,11)

Inchi Key

HXDSPBCJEQJDSB-UHFFFAOYSA-N

Purity

90%

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library