Skip to Content
MilliporeSigma

AABH99A4E21D

AA BLOCKS, INC.

Teneligliptin (hydrobromide hydrate)

Synonym(s):

bis(((2S,4S)-4-(4-(3-methyl-1-phenyl-1H-pyrazol-5-yl)piperazin-1-yl)pyrrolidin-2-yl)(thiazolidin-3-yl)methanone) pentahydrobromide hydrate

Select a Size

Change View

Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/2C22H30N6OS.5BrH.H2O/c2*1-17-13-21(28(24-17)18-5-3-2-4-6-18)26-9-7-25(8-10-26)19-14-20(23-15-19)22(29)27-11-12-30-16-27;;;;;;/h2*2-6,13,19-20,23H,7-12,14-16H2,1H3;5*1H;1H2/t2*19-,20-;;;;;;/m00....../s1

Inchi Key

NQPRXLXVOBOTGT-KSLCDFCZSA-N

Purity

98%

Country of Origin

US

Storage temp.

2~8C





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library